Learn More
Biomatik Corporation CFHR2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508753
Encompass_Preferred
The CFHR2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Complement factor H-related protein 2 protein (20-269AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Complement factor H-related protein 2, FHR-2, CFHR2, CFHL2, FHR2, HFL3, DDESK59, H factor-like 3, H factor-like protein 2. UniProt: P36980. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8753 Supplier Biomatik Corporation Supplier No. CAC09170
The CFHR2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Complement factor H-related protein 2 protein (20-269AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Complement factor H-related protein 2, FHR-2, CFHR2, CFHL2, FHR2, HFL3, DDESK59, H factor-like 3, H factor-like protein 2. UniProt: P36980. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.