Learn More
Biomatik Corporation SAA1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09172
Encompass_Preferred
Saa1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SAA1. The Saa1 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Serum Amyloid A-1 Protein (20-122Aa). Applications: ELISA, IHC. Target Synonyms: SAA1. Accession Number: P0DJI8. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8755