Learn More
Biomatik Corporation ureA anti-Helicobacter pylori, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09174
Encompass_Preferred
Urea Polyclonal Antibody is a Helicobacter pylori Polyclonal Antibody to ureA. The Urea Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Helicobacter pylori. Immunogen: Recombinant Helicobacter Pylori Urease Subunit Alpha Protein (1-238Aa). Applications: ELISA, WB. Target Synonyms: ureA. Accession Number: P14916. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8757