Learn More
Biomatik Corporation ureB anti-Helicobacter pylori, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09175
Encompass_Preferred
Ureb Polyclonal Antibody is a Helicobacter pylori Polyclonal Antibody to ureB. The Ureb Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Helicobacter pylori. Immunogen: Recombinant Helicobacter Pylori Urease Subunit Beta Protein (1-569Aa). Applications: ELISA, WB. Target Synonyms: ureB. Accession Number: P69996. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8758