Learn More
Biomatik Corporation ORM2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09178
Encompass_Preferred
Orm2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ORM2. The Orm2 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Alpha-1-Acid Glycoprotein 2 Protein (19-201Aa). Applications: ELISA, WB. Target Synonyms: ORM2. Accession Number: P19652. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8761