Learn More
Biomatik Corporation HLA-C anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508766
Encompass_Preferred
The HLA-C Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human HLA-C protein (25-308AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: MHC class I antigen, HLA-C, HLA-Cw. UniProt: O78179. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8766 Supplier Biomatik Corporation Supplier No. CAC09183
The HLA-C Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human HLA-C protein (25-308AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: MHC class I antigen, HLA-C, HLA-Cw. UniProt: O78179. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.