Learn More
Biomatik Corporation ATG5 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09184
Encompass_Preferred
The ATG5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Autophagy protein 5 protein (5-272AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Autophagy protein 5, APG5-like, Apoptosis-specific protein, ATG5, APG5L, ASP. UniProt: Q9H1Y0. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8767