Learn More
Biomatik Corporation Avpr1a anti-Rat, Mouse, Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09186
Encompass_Preferred
The Avpr1a Polyclonal Antibody is raised in Rabbit. Species Reactivity: Rat, Mouse, Human. Immunogen: Recombinant rat Avpr1a protein (7-52AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: AVPR V1a Antidiuretic hormone receptor 1a Vascular/hepatic-type arginine vasopressin receptor Avpr1a. UniProt: P30560. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8769