Learn More
Biomatik Corporation HPRT1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508771
Encompass_Preferred
The HPRT1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Hypoxanthine-guanine phosphoribosyltransferase human protein (2-218AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: HGPRT, HPRT1, HPRT. UniProt: P00492. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8771 Supplier Biomatik Corporation Supplier No. CAC09188
The HPRT1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Hypoxanthine-guanine phosphoribosyltransferase human protein (2-218AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: HGPRT, HPRT1, HPRT. UniProt: P00492. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.