Learn More
Biomatik Corporation PRKCA anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508772
Encompass_Preferred
Prkca Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PRKCA. The Prkca Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Protein Kinase C Alpha Type Protein (495-659Aa). Applications: ELISA, IHC, IF. Target Synonyms: PRKCA. Accession Number: P17252. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8772 Supplier Biomatik Corporation Supplier No. CAC09189
Prkca Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PRKCA. The Prkca Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Protein Kinase C Alpha Type Protein (495-659Aa). Applications: ELISA, IHC, IF. Target Synonyms: PRKCA. Accession Number: P17252. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.