Learn More
Biomatik Corporation AKT2 anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09191
Encompass_Preferred
The AKT2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant human RAC-beta serine/threonine-protein kinase protein (1-481AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:100-1:500, IF:1:50-1:500. Alternative Names: Protein kinase Akt-2, Protein kinase B beta, RAC-PK-beta, AKT2. UniProt: P31751. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8774