Learn More
Biomatik Corporation polA anti-Thermus aquaticus, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508777
Encompass_Preferred
Pola Polyclonal Antibody is a Thermus aquaticus Polyclonal Antibody to polA. The Pola Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Thermus aquaticus. Immunogen: Recombinant Thermus Aquaticus Dna Polymerase I, Thermostable Protein (310-832Aa). Applications: ELISA, WB. Target Synonyms: polA. Accession Number: P19821. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8777 Supplier Biomatik Corporation Supplier No. CAC09194
Pola Polyclonal Antibody is a Thermus aquaticus Polyclonal Antibody to polA. The Pola Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Thermus aquaticus. Immunogen: Recombinant Thermus Aquaticus Dna Polymerase I, Thermostable Protein (310-832Aa). Applications: ELISA, WB. Target Synonyms: polA. Accession Number: P19821. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.