Learn More
Biomatik Corporation NEU1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09199
Encompass_Preferred
Neu1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NEU1. The Neu1 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Sialidase-1 Protein (201-415Aa). Applications: ELISA, IHC, IF. Target Synonyms: NEU1. Accession Number: Q99519. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8782