Learn More
Biomatik Corporation MT1X anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09202
Encompass_Preferred
Mt1X Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MT1X. The Mt1X Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Metallothionein-1X Protein (1-59Aa). Applications: ELISA, IHC. Target Synonyms: MT1X. Accession Number: P80297. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8785