Learn More
Biomatik Corporation glpK anti-Escherichia coli, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09209
Encompass_Preferred
The glpK Polyclonal Antibody is raised in Rabbit. Species Reactivity: Escherichia coli. Immunogen: Recombinant Escherichia coli Glycerol kinase protein (2-502AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-1:5000. Alternative Names: ATP:glycerol 3-phosphotransferase, Glycerokinase, glpK. UniProt: P0A6F3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8792