Learn More
Biomatik Corporation BETV4 anti-Betula pendula, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508794
Encompass_Preferred
The BETV4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Betula pendula. Immunogen: Recombinant Betula pendula Polcalcin Bet v 4 protein (1-85AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-1:5000. Alternative Names: Polcalcin Bet v 4, Calcium-binding pollen allergen Bet v 4, Bet v 4, BETV4. UniProt: Q39419. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8794 Supplier Biomatik Corporation Supplier No. CAC09211
The BETV4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Betula pendula. Immunogen: Recombinant Betula pendula Polcalcin Bet v 4 protein (1-85AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-1:5000. Alternative Names: Polcalcin Bet v 4, Calcium-binding pollen allergen Bet v 4, Bet v 4, BETV4. UniProt: Q39419. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.