Learn More
Biomatik Corporation P anti-Rabies virus, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09213
Encompass_Preferred
P Polyclonal Antibody is a Rabies virus Polyclonal Antibody to P. The P Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Rabies virus. Immunogen: Recombinant Rabies Virus Phosphoprotein Protein (1-297Aa). Applications: ELISA, WB. Target Synonyms: P. Accession Number: P69480. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8796