Learn More
Biomatik Corporation C4B anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508797
Encompass_Preferred
The C4B Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Complement C4-B protein (1454-1744AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Basic complement C4, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3, CO4, CPAMD3, C4B, C4B_2. UniProt: P0C0L5. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8797 Supplier Biomatik Corporation Supplier No. CAC09214
The C4B Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Complement C4-B protein (1454-1744AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Basic complement C4, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3, CO4, CPAMD3, C4B, C4B_2. UniProt: P0C0L5. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.