Learn More
Biomatik Corporation pfo anti-Clostridium perfringens, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09215
Encompass_Preferred
Pfo Polyclonal Antibody is a Clostridium perfringens Polyclonal Antibody to pfo. The Pfo Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Clostridium perfringens. Immunogen: Recombinant Clostridium Perfringens Perfringolysin O Protein (29-500Aa). Applications: ELISA, WB. Target Synonyms: pfo. Accession Number: P0C2E9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8798