Learn More
Biomatik Corporation speB anti-Streptococcus pyogenes, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09216
Encompass_Preferred
Speb Polyclonal Antibody is a Streptococcus pyogenes Polyclonal Antibody to speB. The Speb Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Streptococcus pyogenes. Immunogen: Recombinant Streptococcus Pyogenes Streptopain Protein (146-398Aa). Applications: ELISA, WB. Target Synonyms: speB. Accession Number: P0C0J0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8799