Learn More
Biomatik Corporation HLA-C anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508802
Encompass_Preferred
The HLA-C Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human HLA class I histocompatibility antigen, Cw-7 alpha chain protein (62-306AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200. Alternative Names: HLA class I histocompatibility antigen, Cw-7 alpha chain, MHC class I antigen Cw*7, HLA-C, HLAC. UniProt: P10321. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8802 Supplier Biomatik Corporation Supplier No. CAC09219
The HLA-C Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human HLA class I histocompatibility antigen, Cw-7 alpha chain protein (62-306AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200. Alternative Names: HLA class I histocompatibility antigen, Cw-7 alpha chain, MHC class I antigen Cw*7, HLA-C, HLAC. UniProt: P10321. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.