Learn More
Biomatik Corporation ligA anti-Escherichia coli, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09222
Encompass_Preferred
Liga Polyclonal Antibody is a Escherichia coli Polyclonal Antibody to ligA. The Liga Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Escherichia coli. Immunogen: Recombinant Escherichia Coli Dna Ligase Protein (1-671Aa). Applications: ELISA, WB. Target Synonyms: ligA. Accession Number: P15042. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8805