Learn More
Biomatik Corporation cry1Ac anti-Bacillus thuringiensis subsp. Kurstaki, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09226
Encompass_Preferred
The cry1Ac Polyclonal Antibody is raised in Rabbit. Species Reactivity: Bacillus thuringiensis subsp. Kurstaki. Immunogen: Recombinant Bacillus thuringiensis subsp. Kurstaki Pesticidal crystal protein cry1Ac protein (972-1178AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: 133 kDa crystal protein Crystaline entomocidal protoxin cry1Ac cry1A (c), cry218, cryIA (c). UniProt: P05068. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8809