Learn More
Biomatik Corporation tar anti-Escherichia coli, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09227
Encompass_Preferred
Tar Polyclonal Antibody is a Escherichia coli Polyclonal Antibody to tar. The Tar Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Escherichia coli. Immunogen: Recombinant Escherichia Coli Methyl-Accepting Chemotaxis Protein Ii Protein (212-553Aa). Applications: ELISA, WB. Target Synonyms: tar. Accession Number: P07017. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8810