Learn More
Biomatik Corporation surA anti-Escherichia coli, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09230
Encompass_Preferred
Sura Polyclonal Antibody is a Escherichia coli Polyclonal Antibody to surA. The Sura Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Escherichia coli. Immunogen: Recombinant Escherichia Coli Chaperone Sura Protein (21-428Aa). Applications: ELISA, WB. Target Synonyms: surA. Accession Number: P0ABZ6. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8813