Learn More
Biomatik Corporation B4GALT5 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508818
Encompass_Preferred
The B4GALT5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Beta-1,4-galactosyltransferase 5 protein (101-388AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Beta-1, 4-galactosyltransferase 5, Beta-1, 4-GalTase 5, Beta4Gal-T5, b4Gal-T5, Beta-1, 4-GalT II, UDP-Gal:beta-GlcNAc beta-1, 4-galactosyltransferase 5, UDP-galactose:beta-N-acetylglucosamine beta-1, 4-galactosyltransferase 5, B4GALT5. UniProt: O43286. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8818 Supplier Biomatik Corporation Supplier No. CAC09235
The B4GALT5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Beta-1,4-galactosyltransferase 5 protein (101-388AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Beta-1, 4-galactosyltransferase 5, Beta-1, 4-GalTase 5, Beta4Gal-T5, b4Gal-T5, Beta-1, 4-GalT II, UDP-Gal:beta-GlcNAc beta-1, 4-galactosyltransferase 5, UDP-galactose:beta-N-acetylglucosamine beta-1, 4-galactosyltransferase 5, B4GALT5. UniProt: O43286. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.