Learn More
Biomatik Corporation NUPR1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09236
Encompass_Preferred
Nupr1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NUPR1. The Nupr1 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Nuclear Protein 1 Protein (2-64Aa). Applications: ELISA, IHC, IF. Target Synonyms: NUPR1. Accession Number: O60356. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8819