Learn More
Biomatik Corporation luxS anti-Escherichia coli, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09238
Encompass_Preferred
Luxs Polyclonal Antibody is a Escherichia coli Polyclonal Antibody to luxS. The Luxs Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Escherichia coli. Immunogen: Recombinant Escherichia Coli S-Ribosylhomocysteine Lyase Protein (2-171Aa). Applications: ELISA, WB. Target Synonyms: luxS. Accession Number: P45578. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8821