Learn More
Biomatik Corporation IL10RB anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09240
Encompass_Preferred
Il10Rb Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IL10RB. The Il10Rb Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interleukin-10 Receptor Subunit Beta Protein (79-219Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: IL10RB. Accession Number: Q08334. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8823