Learn More
Biomatik Corporation SEMA5A anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09247
Encompass_Preferred
Sema5A Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SEMA5A. The Sema5A Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Semaphorin-5A Protein (201-400Aa). Applications: ELISA, IHC, IF. Target Synonyms: SEMA5A. Accession Number: Q13591. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8830