Learn More
Biomatik Corporation DYNC1H1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09250
Encompass_Preferred
The DYNC1H1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Cytoplasmic dynein 1 heavy chain 1 protein (12-66AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Cytoplasmic dynein 1 heavy chain 1, Cytoplasmic dynein heavy chain 1, Dynein heavy chain, cytosolic, DYNC1H1, DHC1, DNCH1, DNCL, DNECL, DYHC, KIAA0325. UniProt: Q14204. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8833