Learn More
Biomatik Corporation MYBPC3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09252
Encompass_Preferred
Mybpc3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MYBPC3. The Mybpc3 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Myosin-Binding Protein C, Cardiac-Type Protein (1-328Aa). Applications: ELISA, WB. Target Synonyms: MYBPC3. Accession Number: Q14896. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8835