Learn More
Biomatik Corporation TXNRD1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508838
Encompass_Preferred
Txnrd1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TXNRD1. The Txnrd1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Thioredoxin Reductase 1, Cytoplasmic Protein (1-497Aa). Applications: ELISA, WB, IHC. Target Synonyms: TXNRD1. Accession Number: Q16881. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8838 Supplier Biomatik Corporation Supplier No. CAC09255
Txnrd1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TXNRD1. The Txnrd1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Thioredoxin Reductase 1, Cytoplasmic Protein (1-497Aa). Applications: ELISA, WB, IHC. Target Synonyms: TXNRD1. Accession Number: Q16881. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.