Learn More
Biomatik Corporation SQSTM1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09268
Encompass_Preferred
Sqstm1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SQSTM1. The Sqstm1 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Sequestosome-1 Protein (16-310Aa). Applications: ELISA, IHC, IF, IP. Target Synonyms: SQSTM1. Accession Number: Q13501. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8851