Learn More
Biomatik Corporation FAM53B anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09269
Encompass_Preferred
The FAM53B Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Protein FAM53B protein (1-305AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Protein FAM53B, FAM53B, KIAA0140. UniProt: Q14153. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8852