Learn More
Biomatik Corporation RIPK1 anti-Human, Rat, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09275
Encompass_Preferred
Ripk1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RIPK1. The Ripk1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Receptor-Interacting Serine; Threonine-Protein Kinase 1 Protein (328-458Aa). Applications: ELISA, WB, IF. Target Synonyms: RIPK1. Accession Number: Q13546. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8858