Learn More
Biomatik Corporation GPR176 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09278
Encompass_Preferred
The GPR176 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Probable G-protein coupled receptor 176 protein (321-515AA). Applications: ELISA, WB, IF. Recommended dilution: WB:1:1000-1:5000, IF:1:50-1:200. Alternative Names: Probable G-protein coupled receptor 176, HB-954, GPR176. UniProt: Q14439. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8861