Learn More
Biomatik Corporation LY6E anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508866
Encompass_Preferred
Ly6E Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LY6E. The Ly6E Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Lymphocyte Antigen 6E Protein (21-101Aa). Applications: ELISA, IHC, IF. Target Synonyms: LY6E. Accession Number: Q16553. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8866 Supplier Biomatik Corporation Supplier No. CAC09283
Ly6E Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LY6E. The Ly6E Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Lymphocyte Antigen 6E Protein (21-101Aa). Applications: ELISA, IHC, IF. Target Synonyms: LY6E. Accession Number: Q16553. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.