Learn More
Biomatik Corporation TSPAN31 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09295
Encompass_Preferred
Tspan31 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TSPAN31. The Tspan31 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tetraspanin-31 Protein (94-173Aa). Applications: ELISA, IHC, IF. Target Synonyms: TSPAN31. Accession Number: Q12999. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8878