Learn More
Biomatik Corporation MYO7A anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508879
Encompass_Preferred
Myo7A Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MYO7A. The Myo7A Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Unconventional Myosin-Viia Protein (838-968Aa). Applications: ELISA, IHC, IF. Target Synonyms: MYO7A. Accession Number: Q13402. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8879 Supplier Biomatik Corporation Supplier No. CAC09296
Myo7A Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MYO7A. The Myo7A Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Unconventional Myosin-Viia Protein (838-968Aa). Applications: ELISA, IHC, IF. Target Synonyms: MYO7A. Accession Number: Q13402. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.