Learn More
Biomatik Corporation ATR anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09298
Encompass_Preferred
The ATR Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Serine/threonine-protein kinase ATR protein (2405-2644AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Serine/threonine-protein kinase ATR, Ataxia telangiectasia and Rad3-related protein, FRAP-related protein 1, ATR, FRP1. UniProt: Q13535. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8881