Learn More
Biomatik Corporation COTL1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508882
Encompass_Preferred
The COTL1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Coactosin-like protein (2-142AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: COTL1, CLP. UniProt: Q14019. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8882 Supplier Biomatik Corporation Supplier No. CAC09299
The COTL1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Coactosin-like protein (2-142AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: COTL1, CLP. UniProt: Q14019. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.