Learn More
Biomatik Corporation ACAP1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09304
Encompass_Preferred
The ACAP1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 1 protein (1-266AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 1, Centaurin-beta-1, Cnt-b1, ACAP1, CENTB1, KIAA0050. UniProt: Q15027. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8887