Learn More
Biomatik Corporation IL18R1 anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09309
Encompass_Preferred
Il18R1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IL18R1. The Il18R1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interleukin-18 Receptor 1 Protein (22-128Aa). Applications: ELISA, WB, IHC. Target Synonyms: IL18R1. Accession Number: Q13478. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8892