Learn More
Biomatik Corporation EBP anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09315
Encompass_Preferred
The EBP Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Scavenger receptor class B member 1 protein (33-443AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Cholestenol Delta-isomerase, Delta (8)-Delta (7) sterol isomerase, D8-D7 sterol isomerase, Emopamil-binding protein, EBP. UniProt: Q15125. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8898