Learn More
Biomatik Corporation ACKR1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09316
Encompass_Preferred
The ACKR1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Atypical chemokine receptor 1 protein (1-63AA). Applications: ELISA, WB, IF. Recommended dilution: WB:1:1000-1:5000, IF:1:50-1:200. Alternative Names: Atypical chemokine receptor 1, Duffy antigen/chemokine receptor, Glycoprotein D, Plasmodium vivax receptor, ACKR1, DARC, FY, GPD. UniProt: Q16570. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8899