Learn More
Biomatik Corporation dnaK anti-Escherichia coli, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09317
Encompass_Preferred
The dnaK Polyclonal Antibody is raised in Rabbit. Species Reactivity: Escherichia coli. Immunogen: Recombinant Escherichia coli Chaperone protein DnaK protein (1-638AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-1:5000. Alternative Names: Chaperone protein DnaK, HSP70, dnaK. UniProt: Q1RGI8. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8900