Learn More
Biomatik Corporation NGF anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09319
Encompass_Preferred
Ngf Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NGF. The Ngf Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Beta-Nerve Growth Factor Protein (122-218Aa). Applications: ELISA, IHC. Target Synonyms: NGF. Accession Number: P01138. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8902