Learn More
Biomatik Corporation SLC4A8 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508904
Encompass_Preferred
Slc4A8 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC4A8. The Slc4A8 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Electroneutral Sodium Bicarbonate Exchanger 1 Protein (1-300Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SLC4A8. Accession Number: Q2Y0W8. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8904 Supplier Biomatik Corporation Supplier No. CAC09322
Slc4A8 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC4A8. The Slc4A8 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Electroneutral Sodium Bicarbonate Exchanger 1 Protein (1-300Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SLC4A8. Accession Number: Q2Y0W8. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.