Learn More
Biomatik Corporation ANO4 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09324
Encompass_Preferred
The ANO4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Anoctamin-4 protein (1-200AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Anoctamin-4, Transmembrane protein 16D, ANO4, TMEM16D. UniProt: Q32M45. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8906